Learn More
PI3 Kinase p110 Beta Mouse anti-Human, Unconjugated, Clone: 1B2A1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | PI3 Kinase p110 Beta |
|---|---|
| Clone | 1B2A1 |
| Concentration | 860 μg/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16809575
|
Proteintech
67121-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16899565
|
Proteintech
67121-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PI3 Kinase p110 Beta antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PI3 Kinase p110 Beta. This antibody recognizes human, rat antigen. The PI3 Kinase p110 Beta antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| PI3 Kinase p110 Beta | |
| 860 μg/mL | |
| Monoclonal | |
| Mouse | |
| 5291 | |
| Primary |
| 1B2A1 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Human, Rat | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.