Learn More
PI3 Kinase p110 Beta Mouse anti-Human, Unconjugated, Clone: 1B2A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67121-1-IG-20UL
Description
The PI3 Kinase p110 Beta antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PI3 Kinase p110 Beta. This antibody recognizes human, rat antigen. The PI3 Kinase p110 Beta antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| PI3 Kinase p110 Beta | |
| Monoclonal | |
| 860 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat |
| Western Blot, Immunohistochemistry | |
| 1B2A1 | |
| Unconjugated | |
| 20 μL | |
| 5291 | |
| IgG1 |