Learn More
PI3 Kinase p110 Beta Mouse anti-Human, Unconjugated, Clone: 1B2A1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67121-1-IG-150UL
 View more versions of this product
Product Code. 16899565
  • £349.00 / 150µL
Description
The PI3 Kinase p110 Beta antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PI3 Kinase p110 Beta. This antibody recognizes human, rat antigen. The PI3 Kinase p110 Beta antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
PI3 Kinase p110 Beta
Monoclonal
860 μg/mL
Mouse
Primary
Human, Rat
Western Blot, Immunohistochemistry
1B2A1
Unconjugated
150 μL
5291
IgG1