Learn More
PI3 Kinase p110 Alpha Rabbit, anti-Human, Clone: 1B4, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82796-4-RR-100UL
Description
The PI3 Kinase p110 Alpha antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human PI3 Kinase p110 Alpha. This antibody recognizes human, mouse, rat antigen. The PI3 Kinase p110 Alpha antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PI3 Kinase p110 Alpha | |
| 1B4 | |
| p110 alpha, PI3 kinase subunit alpha, PI3K, PI3K alpha, PIK3CA, PtdIns 3 kinase subunit alpha | |
| 100 μL | |
| 5290 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |