Learn More
PI3 Kinase p110 Alpha Rabbit, anti-Human, Clone: 1B4, Proteintech
Rabbit Recombinant Monoclonal Antibody
£135.00 - £360.00
Specifications
| Antigen | PI3 Kinase p110 Alpha |
|---|---|
| Clone | 1B4 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18035385
|
Proteintech
82796-4-RR-100UL |
100 μL |
£360.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18008315
|
Proteintech
82796-4-RR-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PI3 Kinase p110 Alpha antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human PI3 Kinase p110 Alpha. This antibody recognizes human, mouse, rat antigen. The PI3 Kinase p110 Alpha antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PI3 Kinase p110 Alpha | |
| 1000 μg/mL | |
| Liquid | |
| Human, Mouse, Rat | |
| p110 alpha, PI3 kinase subunit alpha, PI3K, PI3K alpha, PIK3CA, PtdIns 3 kinase subunit alpha | |
| Primary |
| 1B4 | |
| Western Blot, ELISA | |
| Rabbit | |
| 5290 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.