missing translation for 'onlineSavingsMsg'
Learn More

PI 3-Kinase p110 delta Antibody (PIK3CD/4639), Novus Biologicals™

Mouse Monoclonal Antibody

£187.00 - £368.00

Specifications

Antigen PI 3-Kinase p110 delta
Clone PIK3CD/4639
Concentration 0.2 mg/mL
Dilution Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18651795
View Documents
Novus Biologicals
NBP3-23783-100ug
100 μg
£368.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18682294
View Documents
Novus Biologicals
NBP3-23783-20ug
20 μg
£187.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PI 3-Kinase p110 delta Monoclonal antibody specifically detects PI 3-Kinase p110 delta in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications

Specifications

PI 3-Kinase p110 delta
0.2 mg/mL
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Autophagy, mTOR Pathway, Phospho Specific, Signal Transduction
10 mM PBS with 0.05% BSA
5293
IgG2b κ
Protein A or G purified
PIK3CD/4639
Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
EC 2.7.1, EC 2.7.1.153, p110D, P110DELTA, Phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta isoform, phosphoinositide-3-kinase C, phosphoinositide-3-kinase, catalytic, delta polypeptide, PI3K, PI3K-delta, PI3-kinase p110 subunit delta, PI3-kinase subunit delta, PtdIns-3-kinase subunit delta, PtdIns-3-kinase subunit p110-delta
Recombinant fragment of human PI 3-Kinase p110 delta protein (around aa 520-720) (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.