missing translation for 'onlineSavingsMsg'
Learn More

PI 3-Kinase p110 delta Antibody (PIK3CD/4639), Novus Biologicals™

Product Code. 18651795 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18651795 100 μg 100µg
18682294 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18651795 Supplier Novus Biologicals Supplier No. NBP323783100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PI 3-Kinase p110 delta Monoclonal antibody specifically detects PI 3-Kinase p110 delta in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PI 3-Kinase p110 delta
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PIK3CD/4639
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Alias EC 2.7.1, EC 2.7.1.153, p110D, P110DELTA, Phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta isoform, phosphoinositide-3-kinase C, phosphoinositide-3-kinase, catalytic, delta polypeptide, PI3K, PI3K-delta, PI3-kinase p110 subunit delta, PI3-kinase subunit delta, PtdIns-3-kinase subunit delta, PtdIns-3-kinase subunit p110-delta
Host Species Mouse
Immunogen Recombinant fragment of human PI 3-Kinase p110 delta protein (around aa 520-720) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, mTOR Pathway, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5293
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.