missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ pHrodo™ Red Phagocytosis Particle Labeling Kit for Flow Cytometry

Product Code. 10761764
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10761764 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10761764 Supplier Invitrogen™ Supplier No. A10026

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

pHrodo™ Red Phagocytosis Particle Labeling Kit for Flow Cytometry

  • Specifically detect phagocytosis and endocytosis with pH-sensitive fluorogenic dye - discriminate endcytosed from adherent and extracellular particles
  • Reduced signal variability and improved timing in sensitive experiments - no need for wash steps or quencher dye
  • Bright red fluorescence - conveniently multiplex with green dyes such as GFP, Fluo-4, or calcein
The pHrodo™ Red dye gives faster and more accurate results than any other phagocytosis assay
The new Molecular Probes™ proprietary pH-sensitive rhodamine-based pHrodo™ Red dye is non-fluorescent at neutral pH, but turns bright red upon acidification. Because it is both fluorogenic and pH-sensitive, the pHrodo™ Red dye can be used as a specific sensor of phagocytic events; acidification of the phagosome following phagocytosis is marked by red fluorescence. It is therefore an ideal tool with which to study phagocytosis and its regulation by drugs and/or environmental factors.

Rapid and reproducible
Wash steps and quencher dyes are not needed since the pHrodo™ Red dye is non-fluorescent outside the cell, making the staining protocol simple and fast. The elimination of wash steps and quencher dyes also improves assay reproducibility, particularly in plate reader-based assays.

Flexible applications
Use ready-made pHrodo™ Red BioParticles™ conjugates for convenient analysis of phagocytosis of Gram-positive or Gram-negative bacteria, or the amine-reactive pHrodo™ Red SE form for labeling microorganisms or proteins of your choice.

Compatible with multiple platforms
pHrodo™ Red dye conjugates can be used in plate readers, fluorescence microscopy imaging, and flow cytometry applications. The pHrodo™ Red Phagocytosis Particle Labeling Kit and the pHrodo™ Red E. coli BioParticles™ Phagocytosis Kit for flow cytometry were designed for rapid and convenient measurements of phagocytic activity in whole blood samples by flow cytometry. The kits include all the reagents required for assessing particle ingestion and red blood cell lysis. The Labeling Kit also contains the reagents required for labeling microorganisms with the pHrodo™ Red dye. Sufficient reagents are provided in both kits for approximately 100 assays.

The optimal absorption and fluorescence emission maxima of the pHrodo™ Red BioParticles™ conjugate is approximately 560nm and 585nm, respectively. However, the fluorophore is readily excited with the 488nm argon-ion laser installed on most flow cytometers.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains lysis buffer (10 mL), buffer B (200 mL), wash buffer (200 mL), 1 vial pHrodo™ red succinimidyl ester (1 mg, lyophilized product), 1 vial DMSO (0.5 mL), 1 bottle sodium bicarbonate (50 mL, 0.1 M, pH 9.3). Store in refrigerator
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometer
Product Type Red Phagocytosis Particle Labeling Kit
Dye Type pHrodo™ Red
Emission 560/585
Format Bottle(s)
Product Line pHrodo
Quantity 1 kit
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.