missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ pHrodo™ Red Microscale Labeling Kit

Product Code. 11622159
Click to view available options
Quantity:
3 Preps
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11622159

Brand: Molecular Probes™ P35363

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

pHrodo™ Red Microscale Labeling Kit

pHrodo™ Red is the new name for pHrodo™, a fluorogenic dye that dramatically increases in fluorescence as the pH of its surroundings become more acidic. Original pHrodo™ and pHrodo™ Red dyes are the same molecule. Use this pHrodo™ Red microscale protein labeling kits to conveniently label 3 separate samples of small amounts (20-100μg) of purified protein or antibody with pHrodo™-SE conjugate.

pHrodo™ Red dye conjugates are non-fluorescent outside the cell, but fluoresce bright red in phagosomes, making them ideal tools for studies ranging from phagocytosis of bioparticles to receptor internalization. pHrodo Red™ offers staining that is faster and more accurate than ratiometric dyes.

The pHrodo™ Red Microscale Labeling Kit Is:
  • Convenient—Labeling typically takes 2 hours with less than 30 min hands-on
  • Versatile—Label proteins with MW between 12 and 150 kDa
  • Customizable—Create your own probes for studying phagocytosis and endocytosis

    Get Better Results in Endocytosis Studies
    Endocytosis and phagocytosis underly physiological events ranging from host-pathogen response to receptor desensitization via internalization (as is common in the GPCR class of receptors). Once internalized, material can be acidified in endosomes or lysosomes. When conjugated to target molecules, our pHrodo™ Red dye fluorogenic pH sensitive probe allows direct visual monitoring of material entering these acidic compartments. At neutral and basic pH values, pHrodo™ Red dye emission is at its minimum; with acidification, pHrodo™ Red dye emission is increased by as much as 8-fold (at pH 4). The fluorescence of pHrodo™ Red dye is dimmed by complete media and whole blood, further improving the signal-to-noise ratio. The pHrodo™ Red Microscale Labeling Kit can label proteins with molecular weights between 12 and 150 kDa, including IgG antibodies (∼150 kDa) in quantities ranging from 20-100μg.

    Gain Experimental Flexibility with a Validated Technology
    pHrodo™ has been cited in over 150 publications and used in a variety of applications. It is validated in flow cytometry, high throughput and high content screening (HTS and HCS), and other imaging applications. pHrodo™ Red offers reduced signal variability and improved timing compared to ratiometric dyes like BCECF and SNARF, and the lack of fluorescence outside the cell eliminates the need for wash steps and quencher dyes. The red fluorescence of pHrodo™ Red is ideal for multiplexing applications with green-fluorescent compounds such GFP, Fluo-4, or calcein.

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Product Type Red Microscale Labeling Kit
    Quantity 3 Preps
    Content And Storage Store at 2°C to 8°C. Desiccate and protect from light.
    For Use With (Equipment) Fluorescence Microscope
    Labeling Scale 20 to 100 μg
    Shipping Condition Room Temperature
    Product Line Molecular Probes™, pHrodo™
    Label or Dye pHrodo Red
    Color Red
    Labeling Target Proteins
    Label Type pH-Sensitive Dyes
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.