missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ pHrodo™ Red Epidermal Growth Factor (EGF) Conjugate
pHrodo™ Red Epidermal Growth Factor (EGF) Conjugate
Brand: Molecular Probes™ P35374
Code : NEW
Additional Details : Weight : 0.00910kg
Description
pHrodo™ Red Epidermal Growth Factor (EGF) Conjugate is used for the detection of internalized EGF receptor in cells. Fluorescently labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more. Fluorescent labels that respond to a pH change add another level of detection. pHrodo™ Red is weakly fluorescent outside of cells at neutral pH, but when internalized specifically through EGF endocytosis, it becomes brightly fluorescent in the acidic endosomes. This allows the measurement of internalized EGF, as well as the dynamic monitoring of EGF internalization in live cells.Features of this EGF conjugate include:
pHrodo™ Red Epidermal Growth Factor (EGF) Conjugate is compatible with various platforms, such as traditional fluorescence microscopy, high content screening (HCS), flow cytometery, and microplate-based fluorimetry or high throughput screening (HTS). This reagent is also compatible with various benchtop instruments such as the Evos™, FLoid™, Tali™, and Attune™ instruments.
Specifications
Orange | |
pHrodo™ Red Epidermal Growth Factor (EGF) Conjugate, 20μg, Orange, Lyophilized, 560/585nm Excitation/Emission, pH-Sensitive Dyes, Used with Flow Cytometer, Fluorescence Microscope, Fluorometer, High Content Instrument | |
Fluorescence Microscopy, High-content Screening (HCS), High-throughput Screening (HTS), Flow Cytometry | |
pHrodo™ Red Epidermal Growth Factor Conjugate | |
560nm |
–5 to –30°C | |
Fluorescent | |
Flow Cytometer, Fluorescence Microscope, Fluorometer, High Content Analysis Instrument | |
pH-Sensitive dyes | |
20μg |
For Research Use Only. Not for use in diagnostic procedures.