missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palintest™ Photometer Kit
This unique photometer kit comes complete in a case with six glass tubes and one dilution tube for the photometer 7100 standard model device.
£1215.00 - £1835.95
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
10676325
|
Palintest™
PTH7100 |
|
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12363263
|
Palintest™
PTBH 7500 |
7500 Model with accessories |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Includes: 7100 Model Standard, with accessories
- Photometer kit for 7100 device
- Comes complete in case with six glass tubes and one dilution tube
- Power supply
- 3 AA batteries
- L x W x H: 146 x 275 x 75 mm
- Weight: 975 g