missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholipase C epsilon 1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | Phospholipase C epsilon 1 |
|---|---|
| Concentration | 2500 μg/mL |
| Applications | Immunohistochemistry, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18049805
|
Proteintech
55463-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18057035
|
Proteintech
55463-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospholipase C epsilon 1 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospholipase C epsilon 1. This antibody recognizes human antigen. The Phospholipase C epsilon 1 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| Phospholipase C epsilon 1 | |
| Immunohistochemistry, ELISA | |
| Rabbit | |
| 51196 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 2500 μg/mL | |
| Liquid | |
| Human | |
| KIAA1516, Phospholipase C epsilon 1, phospholipase C, epsilon 1, PLC epsilon 1, PLCE, PLCE1, PPLC | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts