Learn More
Phospholipase C epsilon 1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55463-1-AP-150UL
Description
The Phospholipase C epsilon 1 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospholipase C epsilon 1. This antibody recognizes human antigen. The Phospholipase C epsilon 1 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| Phospholipase C epsilon 1 | |
| 2500 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Immunohistochemistry, ELISA | |
| KIAA1516, Phospholipase C epsilon 1, phospholipase C, epsilon 1, PLC epsilon 1, PLCE, PLCE1, PPLC | |
| 150 μL | |
| 51196 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |