missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoglucomutase 5 Mouse anti-Human, Mouse, Rat, Bovine, Rabbit, Clone: PGM5/3552, Novus Biologicals™
Mouse Monoclonal Antibody
£197.00 - £396.00
Specifications
| Antigen | Phosphoglucomutase 5 |
|---|---|
| Clone | PGM5/3552 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, ELISA, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18348281
|
Novus Biologicals
NBP3-13996-20UG |
20 μg |
£197.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18306686
|
Novus Biologicals
NBP3-13996-100UG |
100 μg |
£396.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phosphoglucomutase 5 Monoclonal antibody specifically detects Phosphoglucomutase 5 in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| Phosphoglucomutase 5 | |
| 0.2 mg/ml | |
| Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat, Bovine, Rabbit | |
| Q15124 | |
| 5239 | |
| IgG2a κ | |
| Protein A or G purified |
| PGM5/3552 | |
| Western Blot 1-2 ug/ml, ELISA, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| aciculin, EC 5.4.2.2, PGM-RP, PGMRPphosphoglucomutase-like protein 5, phosphoglucomutase 5, Phosphoglucomutase-related protein | |
| Recombinant full-length human Phosphoglucomutase 5 protein (Uniprot: Q15124) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title