missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoglucomutase 5 Mouse anti-Human, Mouse, Rat, Bovine, Rabbit, Clone: PGM5/3552, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-13996-100UG
This item is not returnable.
View return policy
Description
Phosphoglucomutase 5 Monoclonal antibody specifically detects Phosphoglucomutase 5 in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Specifications
| Phosphoglucomutase 5 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 ug/ml, ELISA, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Q15124 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5239 | |
| Store at 4°C. | |
| IgG2a κ |
| Western Blot, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array | |
| PGM5/3552 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| aciculin, EC 5.4.2.2, PGM-RP, PGMRPphosphoglucomutase-like protein 5, phosphoglucomutase 5, Phosphoglucomutase-related protein | |
| Recombinant full-length human Phosphoglucomutase 5 protein (Uniprot: Q15124) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Bovine, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction