missing translation for 'onlineSavingsMsg'
Learn More

Phosphoenolpyruvic acid trisodium salt heptahydrate, 98%

Product Code. 15440287
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15440287

Brand: Thermo Scientific Alfa Aesar J63013.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

When coupled with pyruvate kinase it completes an ATP-regenerating system | CAS: 5541-93-5 | C3H2Na3O6P | 233.986 g/mol

When coupled with pyruvate kinase it completes an ATP-regenerating system

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5541-93-5
Molecular Formula C3H2Na3O6P
Molecular Weight (g/mol) 233.986
MDL Number MFCD00150737
InChI Key RJUFBDMHZGRGMA-UHFFFAOYSA-K
Synonym 2-propenoic acid, 2-phosphonooxy-, trisodium salt, trisodium 2-phosphonatooxy prop-2-enoate, unii-78y3q07j1x, phosphoenolpyruvate trisodium salt, 2-phosphoenol pyruvate na3salt, 2-propenoic acid, 2-phosphonooxy-, sodium salt 1:3, trisodium phosphonatoenolpyruvate, trisodium 2-phosphonatoacrylate, trisodium phosphoenolpyruvate, c3h2o6p.3na
PubChem CID 79673
IUPAC Name trisodium;2-phosphonatooxyprop-2-enoate
SMILES C=C(C(=O)[O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Color White
Quantity 5 g
Formula Weight 360.09
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Phosphoenolpyruvic acid trisodium salt heptahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.