accessibility menu, dialog, popup

UserName

CAS RN 5541-93-5

NaNaNaOOH2COPOOO

CAS RN 5541-93-5

IUPAC Name: trisodium 2-(phosphonatooxy)prop-2-enoate
Synonyms: trisodium phosphonatoenolpyruvate
Molecular Weight (g/mol): 233.99
Molecular Formula: C3H2Na3O6P
InChi Key: RJUFBDMHZGRGMA-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].[O-]C(=O)C(=C)OP([O-])([O-])=O
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results