Learn More
Phospho-STK11/LKB1 (Thr189) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28965-1-AP-20UL
Description
The Phospho-STK11/LKB1 (Thr189) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-STK11/LKB1 (Thr189). This antibody recognizes human, mouse antigen. The Phospho-STK11/LKB1 (Thr189) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Phospho-STK11/LKB1 (Thr189) | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse |
| Western Blot, ELISA | |
| Unconjugated | |
| 20 μL | |
| 6794 | |
| IgG |