Learn More
Phospho-STK11/LKB1 (Thr189) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£125.00 - £280.00
Specifications
| Antigen | Phospho-STK11/LKB1 (Thr189) |
|---|---|
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17442363
|
Proteintech
28965-1-AP-20UL |
20 μL |
£125.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17432363
|
Proteintech
28965-1-AP-100UL |
100 μL |
£280.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-STK11/LKB1 (Thr189) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-STK11/LKB1 (Thr189). This antibody recognizes human, mouse antigen. The Phospho-STK11/LKB1 (Thr189) antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Phospho-STK11/LKB1 (Thr189) | |
| Polyclonal | |
| Rabbit | |
| 6794 | |
| Primary |
| Western Blot, ELISA | |
| Unconjugated | |
| Human, Mouse | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.