missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Stat6 (Tyr641) Recombinant Rabbit Monoclonal Antibody (Stat6Y641-G12), FITC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£185.00 - £465.00
Specifications
| Antigen | Phospho-Stat6 (Tyr641) |
|---|---|
| Clone | Stat6Y641-G12 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17126818
|
Invitrogen™
MA537226 |
10 Tests |
£185.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17156498
![]() |
Invitrogen™
MA536909 |
100 Tests |
£465.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
STAT6 protein is a transcription factor activated by cytokines, particularly interleukin-4 and IL13. (Stat6-/-) were found to be deficient in IL-4-mediated functions including Th2 helper T-cell differentiation, expression of cell surface markers, T-cell proliferation, immunoglobulin class switching to IgE, and partial loss of IL-4 mediated proliferation. STAT6 mRNA has been detected in peripheral blood lymphocytes, colon, intestine, ovary, prostate, thymus, spleen, kidney, liver, lung and placenta. STAT6 is critically involved in Th2 immune response. STAT6 is crucial for IL-4-mediated biological responses, and naturally in two isoforms: STAT6a and STAT6c.Specifications
| Phospho-Stat6 (Tyr641) | |
| Flow Cytometry | |
| FITC | |
| Rabbit | |
| Human, Mouse | |
| P42226, P52633 | |
| 20852, 6778 | |
| A synthetic phospho-peptide corresponding to residues surrounding Tyr641 of human phospho Stat6. | |
| Primary | |
| 4°C, store in dark | |
| STAT6 |
| Stat6Y641-G12 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| 12S1644; D12S1644; IL4 STAT; IL-4 Stat; IL-4-STAT; signal transducer and activator of transcription 6; signal transducer and activator of transcription 6, interleukin-4 induced; signal transducer and transcription activator 6; STAT; STAT, interleukin4-induced; Stat6; STAT-6; STAT6B; STAT6C; transcription factor IL-4 STAT | |
| STAT6 | |
| IgG κ | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title