Learn More
Invitrogen™ Phospho-Stat6 (Tyr641) Recombinant Rabbit Monoclonal Antibody (Stat6Y641-G12), FITC, Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA536909
 View more versions of this product
Product Code. 17156498
  • £465.00 / Pack of 100
Description
STAT6 protein is a transcription factor activated by cytokines, particularly interleukin-4 and IL13. (Stat6-/-) were found to be deficient in IL-4-mediated functions including Th2 helper T-cell differentiation, expression of cell surface markers, T-cell proliferation, immunoglobulin class switching to IgE, and partial loss of IL-4 mediated proliferation. STAT6 mRNA has been detected in peripheral blood lymphocytes, colon, intestine, ovary, prostate, thymus, spleen, kidney, liver, lung and placenta. STAT6 is critically involved in Th2 immune response. STAT6 is crucial for IL-4-mediated biological responses, and naturally in two isoforms: STAT6a and STAT6c.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-Stat6 (Tyr641)
Recombinant Monoclonal
FITC
STAT6
12S1644; D12S1644; IL4 STAT; IL-4 Stat; IL-4-STAT; signal transducer and activator of transcription 6; signal transducer and activator of transcription 6, interleukin-4 induced; signal transducer and transcription activator 6; STAT; STAT, interleukin4-induced; Stat6; STAT-6; STAT6B; STAT6C; transcription factor IL-4 STAT
Rabbit
Protein A/G
RUO
20852, 6778
4°C, store in dark
Liquid
Flow Cytometry
Stat6Y641-G12
PBS with 0.20% BSA and 0.09% sodium azide
P42226, P52633
STAT6
A synthetic phospho-peptide corresponding to residues surrounding Tyr641 of human phospho Stat6.
100 Tests
Primary
Human, Mouse
Antibody
IgG κ