missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-STAT5A (Tyr694) Rabbit anti-Human, FITC, Clone: 2P21, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech FITC-80115-100UL
This item is not returnable.
View return policy
Description
Phospho-STAT5A (Tyr694) antibody apecifically detetects Phospho-STAT5A (Tyr694) antigen in Human Samples. It is validated for Flow Cytometry.Specifications
| Phospho-STAT5A (Tyr694) | |
| 2P21 | |
| FITC | |
| BC027036 | |
| Rabbit | |
| Protein A purification | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| STAT5A | |
| Peptide | |
| 100 μL | |
| 6776 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction