missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-STAT5A (Tyr694) Rabbit anti-Human, FITC, Clone: 2P21, Proteintech
Rabbit Recombinant Antibody
£135.00 - £375.00
Specifications
| Antigen | Phospho-STAT5A (Tyr694) |
|---|---|
| Clone | 2P21 |
| Applications | Flow Cytometry |
| Conjugate | FITC |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17711489
|
Proteintech
FITC-80115-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17650067
|
Proteintech
FITC-80115-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospho-STAT5A (Tyr694) antibody apecifically detetects Phospho-STAT5A (Tyr694) antigen in Human Samples. It is validated for Flow Cytometry.Specifications
| Phospho-STAT5A (Tyr694) | |
| Flow Cytometry | |
| Liquid | |
| Human | |
| BC027036 | |
| STAT5A | |
| IgG | |
| Protein A purification |
| 2P21 | |
| FITC | |
| Rabbit | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 6776 | |
| Peptide | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title