missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-PKA alpha/beta/gamma (Thr197) Recombinant Rabbit Monoclonal Antibody (24GB1650)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen Phospho-PKA alpha/beta/gamma (Thr197)
Clone 24GB1650
Concentration 0.5 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275561
GREENER_CHOICE
View Documents
Invitrogen™
MA556936
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275556
View Documents
Invitrogen™
MA556937
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Phospho-PKA alpha/beta/gamma (Thr197) Recombinant Monoclonal Antibody for Western Blot, Flow

Protein kinase A (PKA, cAMP-dependent protein kinase) is a key element of a ubiquitous signaling pathway important in the cell cycle, cellular communication, memory formation and behavior. PKA is composed of two catalytic (PKAc; Protein Kinase A catalytic subunit) and two regulatory subunits (PKAr). Upon binding cAMP, the complex dissociates to PKAr dimer and two activated PKAc ser/thr protein kinase catalytic monomers. The released PKAc can translocate into the nucleus and exert a regulatory role in the activation of multiple nuclear hormone receptors. However, PKAc-mediated activation of tonicity-dependent gene expression is cAMP independent. Humans express three types of PKAc subunit - PKAc alpha is present in most human tissues, PKAc beta and gamma are tissue-specific, the later is found in testes.
Specifications

Specifications

Phospho-PKA alpha/beta/gamma (Thr197)
0.5 mg/mL
Flow Cytometry, Western Blot
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P05132, P17252, P22612, P22694, P27791, P68181, P68182
18747, 18749, 25636, 293508, 5566, 5567, 5568
A synthesized peptide derived from human Phospho-PKA C (Thr197) (aa 190-204).
Antibody
24GB1650
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
PRKACA
BDPLT19; cAMP-dependent protein kinase catalytic beta subunit isoform 4ab; cAMP-dependent protein kinase catalytic subunit alpha; cAMP-dependent protein kinase catalytic subunit alpha, isoform 1; cAMP-dependent protein kinase catalytic subunit beta; cAMP-dependent protein kinase catalytic subunit gamma; KAPG; PKA C-alpha; PKA C-beta; PKA C-gamma; PKACA; PKACB; PKACg; PPNAD4; PRKACA; PRKACB; PRKACG; protein kinase A catalytic subunit; protein kinase A catalytic subunit beta; protein kinase cAMP-activated catalytic subunit alpha; protein kinase cAMP-activated catalytic subunit beta; protein kinase cAMP-activated catalytic subunit gamma; protein kinase, cAMP-dependent, alpha catalytic subunit; protein kinase, cAMP-dependent, beta catalytic subunit; protein kinase, cAMP-dependent, catalytic, alpha; protein kinase, cAMP-dependent, catalytic, beta; protein kinase, cAMP-dependent, catalytic, gamma; protein kinase, cAMP-dependent, gamma catalytic subunit; serine(threonine) protein kinase
PRKACA, PRKACB, PRKACG
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.