missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-P53 (Ser46) Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
£109.00 - £249.00
Specifications
| Antigen | P53 (Ser46) |
|---|---|
| Clone | 1D10A12 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236589
|
Proteintech
67900-1-IG-100UL |
100 μL |
£280.00 £249.00 / 100µL Save £31.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236829
|
Proteintech
67900-1-IG-20UL |
20 μL |
£125.00 £109.00 / 20µL Save £16.00 12% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-P53 (Ser46) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-P53 (Ser46). This antibody recognizes human antigen. The Phospho-P53 (Ser46) antibody has been validated for the following applications: WB, IHC, IF/ICC, FC (Intra), ELISA analysis.Specifications
| P53 (Ser46) | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 7157 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1D10A12 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| FLJ92943; LFS1; TRP53; p53 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title