missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-P53 (Ser46) Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67900-1-IG-20UL
This item is not returnable.
View return policy
Description
The Phospho-P53 (Ser46) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-P53 (Ser46). This antibody recognizes human antigen. The Phospho-P53 (Ser46) antibody has been validated for the following applications: WB, IHC, IF/ICC, FC (Intra), ELISA analysis.Specifications
| P53 (Ser46) | |
| Monoclonal | |
| 1000 μg/mL | |
| FLJ92943; LFS1; TRP53; p53 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| 53 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| 1D10A12 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 7157 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction