missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-KSR1 (Ser392) Recombinant Rabbit Monoclonal Antibody (KSR1S392-3A4), PE, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537017
This item is not returnable.
View return policy
Description
Part of a multiprotein signaling complex which promotes phosphorylation of Raf family members and activation of downstream MAP kinases. Independently of its kinase activity, acts as MAP2K1/MEK1 and MAP2K2/MEK2-dependent allosteric activator of BRAF; upon binding to MAP2K1/MEK1 or MAP2K2/MEK2, dimerizes with BRAF and promotes BRAF-mediated phosphorylation of MAP2K1/MEK1 and/or MAP2K2/MEK2 (PubMed:29433126). Promotes activation of MAPK1 and/or MAPK3, both in response to EGF and to cAMP. Its kinase activity is unsure. Some protein kinase activity has been detected in vitro, however the physiological relevance of this activity is unknown. [UniProt].
Specifications
| Phospho-KSR1 (Ser392) | |
| Recombinant Monoclonal | |
| PE | |
| KSR1 | |
| AW492498; B-KSR1; CAP kinase; D11Bhm183e; D11Bhm184e; hb; HB protein; kinase KSR1; Kinase suppressor of Ras 1; Ksr; KSR1; mKSR1; OTTHUMP00000162931; protein Hb; RSU2 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 16706, 8844 | |
| 4°C | |
| Liquid |
| Flow Cytometry | |
| KSR1S392-3A4 | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| Q61097, Q8IVT5 | |
| KSR1 | |
| A synthetic phosphor-peptide corresponding to residues surrounding Ser392 of human phospho KSR1. | |
| 100 Tests | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction