Learn More
Phospho-Histone H3 (Ser10) Mouse anti-Human, Unconjugated, Clone: 4C7G2, Proteintech
Mouse Monoclonal Antibody
£125.00 - £280.00
Specifications
| Antigen | Phospho-Histone H3 (Ser10) |
|---|---|
| Clone | 4C7G2 |
| Concentration | 1431 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16849125
|
Proteintech
66863-1-IG-20UL |
20 μL |
£125.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16839125
|
Proteintech
66863-1-IG-100UL |
100 μL |
£280.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-Histone H3 (Ser10) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-Histone H3 (Ser10). This antibody recognizes human, rat, mouse, pig antigen. The Phospho-Histone H3 (Ser10) antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
| Phospho-Histone H3 (Ser10) | |
| 1431 μg/mL | |
| Monoclonal | |
| Mouse | |
| 8350 | |
| Primary |
| 4C7G2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
| Unconjugated | |
| Human, Mouse, Rat, Pig | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.