Learn More
Phospho-Histone H3 (Ser10) Mouse anti-Human, Unconjugated, Clone: 4C7G2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66863-1-IG-100UL
 View more versions of this product
Product Code. 16839125
  • £280.00 / 100µL
Description
The Phospho-Histone H3 (Ser10) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-Histone H3 (Ser10). This antibody recognizes human, rat, mouse, pig antigen. The Phospho-Histone H3 (Ser10) antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
Specifications
Phospho-Histone H3 (Ser10)
Monoclonal
1431 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry
4C7G2
Unconjugated
100 μL
8350
IgG1