missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Histone H2A.X (Ser139) Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
£109.00 - £249.00
Specifications
| Antigen | Histone H2A.X (Ser139) |
|---|---|
| Concentration | 400 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
The Phospho-Histone H2A.X (Ser139) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-Histone H2A.X (Ser139). This antibody recognizes human antigen. The Phospho-Histone H2A.X (Ser139) antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| Histone H2A.X (Ser139) | |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| H2A.X; H2A/X; H2AX | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 400 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 3014 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title