missing translation for 'onlineSavingsMsg'
Learn More

Phospho-Histone H2A.X (Ser139) Rabbit Polyclonal Antibody, Proteintech

Product Code. 30234274 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30234274 100 μL 100µL
30235687 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30234274 Supplier Proteintech Supplier No. 293801AP100UL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The Phospho-Histone H2A.X (Ser139) antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human Phospho-Histone H2A.X (Ser139). This antibody recognizes human antigen. The Phospho-Histone H2A.X (Ser139) antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.

Specifications

Antigen Histone H2A.X (Ser139)
Applications Western Blot, Immunocytochemistry/Immunofluorescence, ELISA
Classification Polyclonal
Concentration 400 μg/mL
Conjugate Unconjugated
Gene Symbols H2A.X; H2A/X; H2AX
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3014
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 15 kDa
Product Type Polyclonal Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.