missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-GSK3B (Ser9) Mouse anti-Human, CoraLite 594, Clone: 1C9E2, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | Phospho-GSK3B (Ser9) |
|---|---|
| Clone | 1C9E2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | CoraLite 594 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17701449
|
Proteintech
CL594-67558-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17619976
|
Proteintech
CL594-67558-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospho-GSK3B (Ser9) Monoclonal antibody apecifically detetects Phospho-GSK3B (Ser9) antigen in Human Samples. It is validated for Flow Cytometry.Specifications
| Phospho-GSK3B (Ser9) | |
| Flow Cytometry | |
| CoraLite 594 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 2932 | |
| Peptide | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
| 1C9E2 | |
| Monoclonal | |
| Liquid | |
| Human | |
| NM_002093 | |
| GSK3B | |
| IgG1 | |
| Protein G purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title