missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-GSK3B (Ser9) Mouse anti-Human, CoraLite 594, Clone: 1C9E2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-67558-100UL
This item is not returnable.
View return policy
Description
Phospho-GSK3B (Ser9) Monoclonal antibody apecifically detetects Phospho-GSK3B (Ser9) antigen in Human Samples. It is validated for Flow Cytometry.Specifications
| Phospho-GSK3B (Ser9) | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| GSK3B | |
| Peptide | |
| 100 μL | |
| 2932 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG1 |
| Flow Cytometry | |
| 1C9E2 | |
| CoraLite 594 | |
| NM_002093 | |
| Mouse | |
| Protein G purification | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction