missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Beta Catenin (Ser675) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28853-1-AP-100UL
This item is not returnable.
View return policy
Description
Phospho-Beta Catenin (Ser675) Polyclonal antibody specifically detects Phospho-Beta Catenin (Ser675) in Human, Mouse samples. It is validated for WB, ELISASpecifications
| Phospho-Beta Catenin (Ser675) | |
| Polyclonal | |
| Unconjugated | |
| BC047565 | |
| Protein A purification | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot, ELISA | |
| 450 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| Rabbit | |
| 100 μL | |
| 1499 | |
| Store at -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction