missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Beta Catenin (Ser675) Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£125.00 - £280.00
Specifications
| Antigen | Phospho-Beta Catenin (Ser675) |
|---|---|
| Concentration | 450 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17781079
|
Proteintech
28853-1-AP-20UL |
20 μL |
£125.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17771079
|
Proteintech
28853-1-AP-100UL |
100 μL |
£280.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospho-Beta Catenin (Ser675) Polyclonal antibody specifically detects Phospho-Beta Catenin (Ser675) in Human, Mouse samples. It is validated for WB, ELISASpecifications
| Phospho-Beta Catenin (Ser675) | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| IgG | |
| Protein A purification | |
| Antibody |
| 450 μg/mL | |
| Polyclonal | |
| Liquid | |
| Human, Mouse | |
| 1499 | |
| Primary | |
| Store at -20°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title