missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Phospho-Akt (S473) Pan Specific DuoSet IC ELISA
Description
Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously.
The Phospho-Akt (S473) Pan Specific DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies phosphorylated human, mouse, rat AKT.
Specifications
Specifications
| Product Type | ELISA Kit |
| Quantity | 1 Pk. (for 15 Plates) |
| Species | Human, Mouse, Rat |
| Conjugate | Biotin |
| Assay Range | 156.00 to 10000 pg/mL |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| Format | 15 96-well Plates |
| For Use With (Application) | ELISA |
| Synonym | RAC-ALPHA, AKT, AKT serine/threonine kinase 1, PKB, PKB alpha, PKB beta, PKB gamma, PKB-ALPHA, PRKBA, RAC, RAC-alpha, RAC-beta, RAC-gamma |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?