missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-4EBP1 (Thr37, Thr46) Recombinant Rabbit Monoclonal Antibody (4EB1T37T46-A5), APC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£185.00 - £464.00
Specifications
| Antigen | Phospho-4EBP1 (Thr37, Thr46) |
|---|---|
| Clone | 4EB1T37T46-A5 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17106838
|
Invitrogen™
MA537244 |
10 Tests |
£185.00
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17146528
![]() |
Invitrogen™
MA536938 |
100 Tests |
£464.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes one member of a family of translation repressor proteins. The protein directly interacts with eukaryotic translation initiation factor 4E, which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.Specifications
| Phospho-4EBP1 (Thr37, Thr46) | |
| Flow Cytometry | |
| APC | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q13541, Q60876, Q62622 | |
| 116636, 13685, 1978 | |
| A synthetic phospho-peptide corresponding to residues surrounding Thr37/46 of human phospho 4E-BP1. | |
| Primary | |
| 4°C | |
| EIF4EBP1 |
| 4EB1T37T46-A5 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| 4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1 | |
| EIF4EBP1 | |
| IgG κ | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title