Learn More
Invitrogen™ Phospho-4EBP1 (Thr37, Thr46) Recombinant Rabbit Monoclonal Antibody (4EB1T37T46-A5), APC, Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA536938
 View more versions of this product
Product Code. 17146528
  • £464.00 / Pack of 100
Description
This gene encodes one member of a family of translation repressor proteins. The protein directly interacts with eukaryotic translation initiation factor 4E, which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-4EBP1 (Thr37, Thr46)
Recombinant Monoclonal
APC
EIF4EBP1
4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1
Rabbit
Protein A/G
RUO
116636, 13685, 1978
4°C
Liquid
Flow Cytometry
4EB1T37T46-A5
PBS with 0.20% BSA and 0.09% sodium azide
Q13541, Q60876, Q62622
EIF4EBP1
A synthetic phospho-peptide corresponding to residues surrounding Thr37/46 of human phospho 4E-BP1.
100 Tests
Primary
Human, Mouse, Rat
Antibody
IgG κ