Learn More
uvex Pheos Eyewear
Product Code. 17757689 Shop All Uvex Products
Change view
Click to view available options
Frame Color: Black and gray ; Coating: supravision ETC
Product Code.
17757689 Frame Color: Black and gray ; Coating: supravision ETC
1 options
Product Code. 17757689 Supplier uvex Supplier No. 9192415
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
More than just a common spectacle - the modern fashion-look makes it a welcome companion at everyday work
  • Perfect all-round protection, large field of vision through duo spherical lens, outstanding ventilation thanks to integrated uvex climazone temple concept
  • Coating uvex supravision excellence
  • TRUSTED_SUSTAINABILITY
    Specifications
    Lens Type Supravision ETC
    Lens Coating Anti-fog
    Color Anthracite, Blue
    Certifications/Compliance (Europe) EN 166, EN 170
    Lens Tint Clear
    Product Type Ergonomic Safety Glasses
    Material Polycarbonate
    Disposable Reusable
    Lens Material Polycarbonate
    Frame Material Plastic
    UV Protection UV400
    Frame Type Standard