Learn More
uvex Pheos S Spectacles
Product Code. 31910427 Shop All Uvex Products
Change view
Click to view available options
Lens Tint:
Gray
Unit Size:
Each
Product Code. Lens Tint unitSize
31910427 Gray Each
1 options
Product Code. 31910427 Supplier uvex Supplier No. 9192786
  • ONLINE EXCLUSIVE PRICE
    £12.66
    £11.63 / Pack of 1
    Save £1.03
    8% Off
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
The uvex pheos s are not just safety spectacles: Their modern, fashionable look makes them a welcome addition to the working day.
The attractive design is underpinned by innovative safety eyewear technology: The main feature of the spectacles is the duo-spherical lens.This lens not only offers a wide field of vision, but it is also particularly scratch-resistant on the outside and features permanent anti-fog protection on the inside thanks to high-quality uvex supra-vision coating technology. By adding an extra frame and headband, the uvex pheos can be expanded into a set, depending on the field of use.
  • Sporty, timeless safety spectacles
  • Frameless design affords an extra-large field of vision
  • Robust polycarbonate lens with proven uvex supra-vision coating technology - permanently anti-fog and scratch-resistant
  • Optional extra frame offers additional protection against particles, dirt and dust
  • Optional headband keeps the spectacles in position during demanding work
  • Narrow version, also available in standard size
  • Fits uvex IES Metal-free
  • Ergonomically shaped side arms with soft ear-pieces ensure a comfortable, secure fit without creating pressure points
  • Anatomically shaped nose piece for all-day wearer comfort
  • Certified in accordance with EN 166 (personal eye protection) and EN 172 (sunglare filters for industrial use) UV400 - 100% protection against harmful UV radiation to 400 nm
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant, Anti-scratch
Lens Tint Gray
Product Type Safety Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material TPE
UV Protection UV400
Frame Type Semi-rimless