Learn More
uvex Pheos CX2 Glasses
Product Code. 31910426 Shop All Uvex Products
Change view
Click to view available options
Lens Tint:
Amber
Clear
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint unitSize
31910426 Clear Each
31910423 Amber Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31910426 Supplier uvex Supplier No. 9198376
  • ONLINE EXCLUSIVE PRICE
    £13.11
    £12.03 / Pack of 1
    Save £1.08
    8% Off
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
Optimal eye protection for a wide range of applications: The soft brow-guard component is injection moulded onto the lens and keeps dust and liquid splashes out of the eyes. The innovative uvex x-tended eyeshield adapts perfectly to the wearer - for high levels of comfort and protection.
  • Sporty safety spectacles with high wearer acceptance
  • Robust polycarbonate lens with proven uvex supra-vision coating technology - permanently anti-fog and scratch-resistant
  • Metal-free
  • Comfort features X-tended eyeshield: Soft-seal frame technology wraps around the forehead and provides protection against particle penetration
  • Ergonomically shaped side arms with soft ear-pieces ensure a comfortable and secure fit without creating pressure points
  • Soft, flexible nose piece adapts to the wearer to deliver a non-slip, pressure-free fit
  • Certified in accordance with EN 166 (personal eye protection) and EN 170 (UV filters) UV400 -100% protection against harmful UV radiation up to 400 nm
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Anti-scratch, Scratch-resistant
Lens Tint Clear
Product Type Safety Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material TPE
UV Protection UV400
Frame Type Semi-rimless