missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenyl nicotinate, 99%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
In nicotinic acid analogs assayed against tri-o-cresyl phosphate-induced ataxia in hens Phenyl nicotinate provided partial relief of the syndrome.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
In nicotinic acid analogs assayed against tri-o-cresyl phosphate-induced ataxia in hens Phenyl nicotinate provided partial relief of the syndrome.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | 70°C to 74°C |
Boiling Point | 186°C to 188°C (20 mmHg) |
Quantity | 5 g |
Beilstein | 140968 |
Formula Weight | 199.21 |
Percent Purity | 99% |
Chemical Name or Material | Phenyl nicotinate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'