missing translation for 'onlineSavingsMsg'
Learn More

Phenyl nicotinate, 99%, Thermo Scientific™

Catalog Number 11405856
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
3468-53-9
C12H9NO2
199.209
MFCD00023581
QEJPOSAIULNDLU-UHFFFAOYSA-N
phenyl nicotinate, 3-pyridinecarboxylic acid, phenyl ester, phenylnicotinate, phenyl 3-pyridinecarboxylate, nicotinic acid, phenyl ester, unii-vrd2l6rm16, vrd2l6rm16, acmc-209i9k, nicotinic acid phenyl ester, qejposaiulndlu-uhfffaoysa
77018
phenyl pyridine-3-carboxylate
C1=CC=C(C=C1)OC(=O)C2=CN=CC=C2
This item is not returnable. View return policy
3468-53-9
C12H9NO2
199.209
MFCD00023581
QEJPOSAIULNDLU-UHFFFAOYSA-N
phenyl nicotinate, 3-pyridinecarboxylic acid, phenyl ester, phenylnicotinate, phenyl 3-pyridinecarboxylate, nicotinic acid, phenyl ester, unii-vrd2l6rm16, vrd2l6rm16, acmc-209i9k, nicotinic acid phenyl ester, qejposaiulndlu-uhfffaoysa
77018
phenyl pyridine-3-carboxylate
C1=CC=C(C=C1)OC(=O)C2=CN=CC=C2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
In nicotinic acid analogs assayed against tri-o-cresyl phosphate-induced ataxia in hens Phenyl nicotinate provided partial relief of the syndrome.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 70°C to 74°C
Boiling Point 186°C to 188°C (20 mmHg)
Quantity 5 g
Beilstein 140968
Formula Weight 199.21
Percent Purity 99%
Chemical Name or Material Phenyl nicotinate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'