missing translation for 'onlineSavingsMsg'
Learn More

Phenyl 2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-Alpha-D-glucopyranoside, TRC

Product Code. 30411916
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30411916 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411916 Supplier Toronto Research Chemicals Supplier No. TRCP30625050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13089-19-5
Molecular Formula C20 H25 N O9
Molecular Weight (g/mol) 423.41
IUPAC Name [(2R,3S,4R,5R,6R)-5-acetamido-3,4-diacetyloxy-6-phenoxyoxan-2-yl]methyl acetate
SMILES CC(=O)N[C@H]1[C@@H](Oc2ccccc2)O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C20H25NO9/c1-11(22)21-17-19(28-14(4)25)18(27-13(3)24)16(10-26-12(2)23)30-20(17)29-15-8-6-5-7-9-15/h5-9,16-20H,10H2,1-4H3,(H,21,22)/t16-,17-,18-,19-,20+/m1/s1
Formula Weight 423.1529
Chemical Name or Material Phenyl 2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-a-D-glucopyran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.