CAS RN 13089-19-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Phenyl 2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-Alpha-D-glucopyranoside, TRC
CAS: 13089-19-5 Molecular Formula: C20 H25 N O9 Molecular Weight (g/mol): 423.41 IUPAC Name: [(2R,3S,4R,5R,6R)-5-acetamido-3,4-diacetyloxy-6-phenoxyoxan-2-yl]methyl acetate SMILES: CC(=O)N[C@H]1[C@@H](Oc2ccccc2)O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C