missing translation for 'onlineSavingsMsg'
Learn More

Phenol Rutinoside, TRC

Product Code. 30138095
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138095 10 mg 10mg
30137537 5 mg 5mg
30009690 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138095 Supplier Toronto Research Chemicals Supplier No. TRCP71565010MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1416253-67-2
Molecular Formula C18 H26 O10
Molecular Weight (g/mol) 402.393
Synonym Phenyl 6-O-(6-deoxy-a-L-mannopyranosyl)-β -D-glucopyranoside
IUPAC Name (2S,3R,4R,5R,6R)-2-methyl-6-[[(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-phenoxyoxan-2-yl]methoxy]oxane-3,4,5-triol
SMILES C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](Oc3ccccc3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H26O10/c1-8-11(19)13(21)15(23)17(26-8)25-7-10-12(20)14(22)16(24)18(28-10)27-9-5-3-2-4-6-9/h2-6,8,10-24H,7H2,1H3/t8-,10+,11-,12+,13+,14-,15+,16+,17+,18+/m0/s1
Formula Weight 402.1526
Chemical Name or Material Phenol Rutinoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.