missing translation for 'onlineSavingsMsg'
Learn More

PHCA Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£328.00 - £513.00

Specifications

Antigen PHCA
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18658416
View Documents
Novus Biologicals
NBP2-68894-25ul
25 μL
£328.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18676687
View Documents
Novus Biologicals
NBP2-68894
100 μg
£513.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PHCA Polyclonal antibody specifically detects PHCA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

PHCA
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
Alkaline CDase 3, alkaline ceramidase 3, Alkaline dihydroceramidase SB89, Alkaline phytoceramidase, aPHC, APHCAlkCDase 3, EC 3.5.1.-, FLJ11238, PHCA, phytoceramidase, alkaline
This antibody was developed against a recombinant protein corresponding to amino acids: DREGYWGPTTSTLDWCEENYSVTWYIAEFWNTVSNLIMIIPPMFGAVQSVRDGLEKR
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Purified
RUO
PBS (pH 7.2) and 40% Glycerol
55331
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.