missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHCA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68894
This item is not returnable.
View return policy
Description
PHCA Polyclonal antibody specifically detects PHCA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PHCA | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Alkaline CDase 3, alkaline ceramidase 3, Alkaline dihydroceramidase SB89, Alkaline phytoceramidase, aPHC, APHCAlkCDase 3, EC 3.5.1.-, FLJ11238, PHCA, phytoceramidase, alkaline | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DREGYWGPTTSTLDWCEENYSVTWYIAEFWNTVSNLIMIIPPMFGAVQSVRDGLEKR | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 55331 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction