Learn More
Agilent BioTek Phase Contrast Objectives for Imaging Multi-Mode Reader
GREENER_CHOICE
Product Code. 15665687 Shop All Agilent BioTek Products
Click to view available options
Product Type:
10X Phase Contrast Objective
20X Phase Contrast Objective
40X Phase Contrast Objective
4X Phase Contrast Objective
Unit Size:
Each
Product Code. 15665687
Brand: Agilent BioTek 1320515
For use with Cytation 5 Cell Imaging Multi-Mode Readers.
Available with magnification of 4X, 10X, 20X and 40X.
TRUSTED_SUSTAINABILITY
Specifications
Product Type 4X Phase Contrast Objective
For Use With (Equipment) Cytation 5 Cell Imaging Multi-Mode Reader