Learn More
Invitrogen™ eBioscience™ Phalloidin eFluor™ 520
Description
Phalloidin is a peptide toxin isolated from the Amanita phalloides mushroom. Phalloidin selectively binds F-actin and functions to stablilize actin polymers by inhibiting the dissociation of actin monomers from the filament ends. Because F-actin polymers compose the cytoskeleton of almost all eukaryotic cells, phalloidin conjugates can be useful probes for visualizing the structure of live or fixed cells using microscopy. Phalloidin eFluor 520 can be used to label cells of multiple species. Its exitation and emission is similar to Alexa Fluor 488 and FITC.
Phalloidin eFluor 520 is supplied as a frozen liquid. To prepare the 1000X stock solution, thaw the vial, add 30μL high-quality anhydrous DMSO, mix and briefly spin down. The resulting solution (phalloidin DMSO stock) should be aliquoted, protected from light and moisture, and stored at -20°C. To prepare a 1X working solution of phalloidin conjugate, add 1μL of 1000X phalloidin DMSO stock solution to 1mL of PBS containing 1% BSA.
Specifications
Specifications
| Excitation Wavelength Range | 488/520 |
| Dye Type | eFluor 520 |
| No. of Tests | 300 tests |
| Quantity | 300 tests |
| Product Line | eBioscience |
| Label Type | eFluor |
| Product Type | Phalloidin |
| SubCellular Localization | Cytoskeleton |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.